siRNA Details
| Gene Symbol: LCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122769.2 | NP_001116241.1 | 3 | 940 | ||
| NM_181714.3 | NP_859065.2 | 4 | 1095 | |||
| XM_005248665.4 | XP_005248722.1 | 3 | 954 | |||
| XM_011535504.1 | XP_011533806.1 | 4 | 1007 | |||
| GenBank mRNA | AK299314.1 | - | 3 | 911 | ||
| BC050327.1 | - | 4 | 1095 | 4823983 | ||
| BX648161.1 | - | 3 | 921 | |||

LCA5
Genomic Map