siRNA Details
| Gene Symbol: LCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122769.2 | NP_001116241.1 | 8 | 2217 | ||
| NM_181714.3 | NP_859065.2 | 9 | 2372 | |||
| XM_005248665.4 | XP_005248722.1 | 8 | 2231 | |||
| XM_011535504.1 | XP_011533806.1 | 9 | 2284 | |||
| GenBank mRNA | AL832214.1 | - | 2 | 676 | ||
| BC050327.1 | - | 9 | 2372 | 4823983 | ||
| BX648161.1 | - | 8 | 2198 | |||

LCA5
Genomic Map