siRNA Details
| Gene Symbol: OR2L8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001963.1 | NP_001001963.1 | 1 | 177 | ||
| GenBank mRNA | BC140751.1 | - | 1 | 258 | 9021619 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2L8| Gene Symbol: OR2L8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001963.1 | NP_001001963.1 | 1 | 177 | ||
| GenBank mRNA | BC140751.1 | - | 1 | 258 | 9021619 | |