siRNA Details
| Gene Symbol: OR2T33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004695.1 | NP_001004695.1 | 1 | 803 | ||
| GenBank mRNA | BC136994.1 | - | 1 | 825 | 9020986 | |
| BC136995.1 | - | 1 | 825 | 9020987 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2T33| Gene Symbol: OR2T33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004695.1 | NP_001004695.1 | 1 | 803 | ||
| GenBank mRNA | BC136994.1 | - | 1 | 825 | 9020986 | |
| BC136995.1 | - | 1 | 825 | 9020987 | ||