siRNA Details
| Gene Symbol: SPINT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178455.1 | NP_848550.1 | 2 | 273 | ||
| GenBank mRNA | AY372173.1 | - | 2 | 273 | ||
| AY372174.1 | - | 2 | 273 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPINT4| Gene Symbol: SPINT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178455.1 | NP_848550.1 | 2 | 273 | ||
| GenBank mRNA | AY372173.1 | - | 2 | 273 | ||
| AY372174.1 | - | 2 | 273 | |||