siRNA Details
| Gene Symbol: OR13C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004481.1 | NP_001004481.1 | 1 | 517 | ||
| GenBank mRNA | BC136824.1 | - | 1 | 560 | 9020815 | |
| BC136826.1 | - | 1 | 560 | 9020817 | ||
| Gene Symbol: OR13C5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004482.1 | NP_001004482.1 | 1 | 517 | ||
| GenBank mRNA | BC136842.1 | - | 1 | 611 | 9020833 | |
| BC136843.1 | - | 1 | 611 | 9020834 | ||

OR13C2
Genomic Map