siRNA Details
| Gene Symbol: OR5C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001923.1 | NP_001001923.1 | 1 | 404 | ||
| GenBank mRNA | BC136700.1 | - | 1 | 466 | 9020690 | |
| BC136701.1 | - | 1 | 466 | 9020691 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5C1| Gene Symbol: OR5C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001923.1 | NP_001001923.1 | 1 | 404 | ||
| GenBank mRNA | BC136700.1 | - | 1 | 466 | 9020690 | |
| BC136701.1 | - | 1 | 466 | 9020691 | ||