siRNA Details
| Gene Symbol: HMCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291815.1 | NP_001278744.1 | 21, 22 | 3202 | ||
| XM_011518465.2 | XP_011516767.1 | 21, 22 | 3339 | |||
| XM_011518466.2 | XP_011516768.1 | 21, 22 | 3339 | |||
| XM_011518467.2 | XP_011516769.1 | 21, 22 | 3339 | |||
| XM_011518468.2 | XP_011516770.1 | 21, 22 | 3339 | |||
| XM_011518469.2 | XP_011516771.1 | 21, 22 | 3339 | |||
| XM_011518470.2 | XP_011516772.1 | 21, 22 | 3339 | |||
| GenBank mRNA | AK128020.1 | - | 9, 10 | 1162 | ||
| BC146865.1 | - | 8, 9 | 1050 | 9056793 | ||

HMCN2
Genomic Map