siRNA Details
| Gene Symbol: GLOD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080489.2 | NP_001073958.2 | 3 | 352 | ||
| XM_011543911.2 | XP_011542213.1 | 4 | 455 | |||
| XM_011543912.2 | XP_011542214.1 | 4 | 599 | |||
| GenBank mRNA | DY655159.1 | - | 2 | 193 | ||

GLOD5
Genomic Map