siRNA Details
| Gene Symbol: TBK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013254.3 | NP_037386.1 | 6 | 845 | ||
| XM_005268809.1 | XP_005268866.1 | 6 | 923 | |||
| XM_005268810.1 | XP_005268867.1 | 6 | 905 | |||
| GenBank mRNA | AF174536.1 | - | 5 | 686 | ||
| AF191838.1 | - | 6 | 748 | |||
| AK291039.1 | - | 6 | 790 | |||
| AK303683.1 | - | 1 | 62 | |||
| BC034950.2 | - | 6 | 785 | 4824988 | ||

TBK1
Genomic Map