siRNA Details
| Gene Symbol: C9orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256054.2 | NP_001242983.1 | 4, 5 | 807 | ||
| NM_018325.4 | NP_060795.1 | 4, 5 | 712 | |||
| NM_145005.6 | NP_659442.2 | 4, 5 | 729 | |||
| GenBank mRNA | AK057806.1 | - | 4, 5 | 707 | ||
| AK291425.1 | - | 4, 5 | 702 | |||
| AL832229.1 | - | 3, 4 | 669 | |||
| AL832467.1 | - | 4, 5 | 668 | |||
| BC020851.2 | - | 3, 4 | 649 | |||
| BC068445.1 | - | 3, 4 | 661 | 5298741 | ||
| JN681271.1 | - | 4, 5 | 785 | |||

C9orf72
Genomic Map