siRNA Details
| Gene Symbol: PSEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000021.3 | NP_000012.1 | 12 | 1653 | ||
| NM_007318.2 | NP_015557.2 | 12 | 1641 | |||
| XM_005267864.2 | XP_005267921.1 | 12 | 1647 | |||
| XM_005267866.1 | XP_005267923.1 | 12 | 1636 | |||
| XM_011536971.2 | XP_011535273.1 | 12 | 1545 | |||
| XM_011536972.2 | XP_011535274.1 | 12 | 1909 | |||
| XM_011536973.1 | XP_011535275.1 | 12 | 1897 | |||
| XM_011536974.1 | XP_011535276.1 | 12 | 1538 | |||
| GenBank mRNA | AK122722.1 | - | 10 | 2922 | ||
| AK223173.1 | - | 10 | 1383 | |||
| AK303876.1 | - | 8 | 1028 | |||
| AK312531.1 | - | 12 | 1637 | |||
| BC011729.2 | - | 12 | 1622 | 4423098 | ||
| KU178285.1 | - | 10 | 1357 | |||
| KU178286.1 | - | 10 | 1369 | |||
| KU178287.1 | - | 5 | 588 | |||
| L42110.1 | - | 12 | 1617 | |||
| L76517.1 | - | 12 | 1910 | |||
| U40379.1 | - | 10 | 1357 | |||
| U40380.1 | - | 11 | 1449 | |||

PSEN1
Genomic Map