siRNA Details
| Gene Symbol: GALC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000153.3 | NP_000144.2 | 12 | 1379 | ||
| NM_001201401.1 | NP_001188330.1 | 11 | 1310 | |||
| NM_001201402.1 | NP_001188331.1 | 12 | 1360 | |||
| XM_011536618.2 | XP_011534920.1 | 12 | 1661 | |||
| GenBank mRNA | AK294040.1 | - | 13 | 1224 | ||
| AK296530.1 | - | 11 | 1201 | |||
| AK302519.1 | - | 7 | 795 | |||
| AK302683.1 | - | 12 | 1533 | |||
| BC036518.2 | - | 12 | 1231 | 5267705 | ||
| D25283.1 | - | 12 | 1262 | |||
| L23116.1 | - | 12 | 1271 | |||

GALC
Genomic Map