siRNA Details
| Gene Symbol: PACRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006715344.3 | XP_006715407.1 | 4, 5 | 966 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PACRG| Gene Symbol: PACRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006715344.3 | XP_006715407.1 | 4, 5 | 966 | ||