siRNA Details
| Gene Symbol: CFHR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030787.3 | NP_110414.1 | 8 | 1341 | ||
| XM_011510020.2 | XP_011508322.1 | 8 | 1248 | |||
| GenBank mRNA | AF295327.1 | - | 8 | 1306 | ||
| AK314026.1 | - | 8 | 1342 | |||
| BC026282.1 | - | 8 | 1322 | |||
| BC111773.1 | - | 8 | 1306 | 40034462 | ||

CFHR5
Genomic Map