siRNA Details
| Gene Symbol: AGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000027.3 | NP_000018.2 | 6 | 763 | ||
| NM_001171988.1 | NP_001165459.1 | 6 | 763 | |||
| GenBank mRNA | AK312982.1 | - | 6 | 697 | ||
| BC012392.1 | - | 6 | 686 | 3859317 | ||
| CR541715.1 | - | 6 | 635 | |||
| M64073.1 | - | 6 | 638 | |||
| M64075.1 | - | 4 | 331 | |||
| X55762.1 | - | 6 | 662 | |||

AGA
Genomic Map