siRNA Details
| Gene Symbol: AGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000027.3 | NP_000018.2 | 5 | 705 | ||
| NM_001171988.1 | NP_001165459.1 | 5 | 705 | |||
| NR_033655.1 | 5 | 705 | ||||
| GenBank mRNA | AK303370.1 | - | 4 | 498 | ||
| AK312982.1 | - | 5 | 639 | |||
| BC012392.1 | - | 5 | 628 | 3859317 | ||
| CR541715.1 | - | 5 | 577 | |||
| CX869892.1 | - | 5 | 582 | |||
| M64073.1 | - | 5 | 580 | |||
| M64075.1 | - | 3 | 273 | |||
| M64076.1 | - | 3 | 273 | |||
| X55762.1 | - | 5 | 604 | |||

AGA
Genomic Map