siRNA Details
| Gene Symbol: SPRED3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042522.2 | NP_001035987.1 | 2 | 332 | ||
| NR_073032.1 | 2 | 147 | ||||
| XM_006723219.3 | XP_006723282.1 | 3 | 323 | |||
| XM_011526978.2 | XP_011525280.1 | 2 | 378 | |||
| GenBank mRNA | BC151136.1 | - | 2 | 332 | 9057036 | |
| BF983829.1 | - | 2 | 149 | |||
| DQ323927.1 | - | 2 | 233 | |||
| DQ323928.1 | - | 2 | 233 | |||

SPRED3
Genomic Map