siRNA Details
| Gene Symbol: NDUFA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193375.1 | NP_001180304.1 | 4 | 953 | ||
| GenBank mRNA | AK127692.1 | - | 4 | 726 | ||
| BQ955016.1 | - | 4 | 699 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDUFA11| Gene Symbol: NDUFA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193375.1 | NP_001180304.1 | 4 | 953 | ||
| GenBank mRNA | AK127692.1 | - | 4 | 726 | ||
| BQ955016.1 | - | 4 | 699 | |||