siRNA Details
| Gene Symbol: ARID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152641.2 | NP_689854.2 | 15 | 3879 | ||
| XM_006719272.3 | XP_006719335.1 | 15 | 4033 | |||
| XM_011538025.1 | XP_011536327.1 | 9 | 2986 | |||
| GenBank mRNA | AB046777.1 | - | 1 | 792 | ||
| AK027718.1 | - | 1 | 1033 | |||
| AK075385.1 | - | 1 | 433 | |||
| AK127872.1 | - | 6 | 1700 | |||
| AL832200.1 | - | 11 | 3432 | |||
| BC090062.1 | - | 1 | 954 | |||
| BX648152.1 | - | 9 | 3167 | |||
| BX649133.1 | - | 7 | 2725 | |||
| CR749833.1 | - | 14 | 3798 | |||
| DQ096628.1 | - | 15 | 3879 | |||

ARID2
Genomic Map