siRNA Details
| Gene Symbol: GDF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001557.2 | NP_001001557.1 | 2 | 567 | ||
| GenBank mRNA | AJ537424.1 | - | 2 | 567 | ||
| BC127947.1 | - | 2 | 494 | 40132943 | ||
| BC127948.1 | - | 2 | 491 | 40132947 | ||

GDF6
Genomic Map