siRNA Details
| Gene Symbol: MEX3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174118.1 | NP_001167589.1 | 3 | 2332 | ||
| NM_203304.3 | NP_976049.3 | 2 | 2519 | |||
| XM_017026811.1 | XP_016882300.1 | 2 | 2319 | |||
| GenBank mRNA | AB062435.1 | - | 1 | 537 | ||
| AB107353.1 | - | 2 | 2112 | |||
| AF458084.1 | - | 3 | 1804 | |||

MEX3D
Genomic Map