siRNA Details
| Gene Symbol: FMOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002023.4 | NP_002014.2 | 2 | 520 | ||
| GenBank mRNA | AK092899.1 | - | 2 | 190 | ||
| AK172740.1 | - | 213 | ||||
| AK291632.1 | - | 2 | 186 | |||
| AK303866.1 | - | 2 | 186 | |||
| AL542756.3 | - | 2 | 583 | |||
| BC035281.1 | - | 2 | 197 | 5181410 | ||
| DC296345.1 | - | 2 | 521 | |||
| X75546.1 | - | 1 | 136 | |||

FMOD
Genomic Map