siRNA Details
| Gene Symbol: PEX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000287.3 | NP_000278.3 | 13 | 2506 | ||
| NM_001316313.1 | NP_001303242.1 | 13 | 2242 | |||
| NR_133009.1 | 11 | 2259 | ||||
| XM_011514661.1 | XP_011512963.1 | 12 | 2422 | |||
| GenBank mRNA | AB051076.1 | - | 13 | 2413 | ||
| AB051077.1 | - | 13 | 2149 | |||
| AB051078.1 | - | 11 | 2166 | |||
| AB208906.1 | - | 11 | 2856 | |||
| AK314237.1 | - | 13 | 2442 | |||
| BC033487.1 | - | 13 | 2421 | |||
| BC048331.1 | - | 13 | 2429 | 5189685 | ||
| D83703.1 | - | 13 | 2483 | |||

PEX6
Genomic Map