siRNA Details
| Gene Symbol: CSF3R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000760.3 | NP_000751.1 | 17 | 3272 | ||
| NM_156039.3 | NP_724781.1 | 17 | 3353 | |||
| NM_172313.2 | NP_758519.1 | 18 | 2852 | |||
| XM_005270493.1 | XP_005270550.1 | 17 | 3269 | |||
| XM_011540748.2 | XP_011539050.1 | 16 | 2878 | |||
| XM_011540749.1 | XP_011539051.1 | 17 | 3350 | |||
| XM_011540750.1 | XP_011539052.1 | 12 | 2183 | |||
| XM_017000370.1 | XP_016855859.1 | 16 | 2915 | |||
| GenBank mRNA | AB209656.1 | - | 9 | 5938 | ||
| BC053585.1 | - | 17 | 2865 | 5217305 | ||
| M59818.1 | - | 17 | 2857 | |||
| M59819.1 | - | 17 | 2769 | |||
| M59820.1 | - | 17 | 2938 | |||
| X55720.1 | - | 18 | 2432 | |||
| X55721.1 | - | 17 | 2853 | |||

CSF3R
Genomic Map