siRNA Details
| Gene Symbol: CYP3A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000777.4 | NP_000768.1 | 13 | 1664 | ||
| NM_001291829.1 | NP_001278758.1 | 14 | 2164 | |||
| NM_001291830.1 | NP_001278759.1 | 14 | 1924 | |||
| NR_033807.2 | 13 | 3268 | ||||
| XM_011515843.1 | XP_011514145.1 | 14 | 1781 | |||
| XM_017011781.1 | XP_016867270.1 | 10 | 1306 | |||
| XM_017011782.1 | XP_016867271.1 | 8 | 1103 | |||
| GenBank mRNA | AA076026.1 | - | 2 | 251 | ||
| AK223008.1 | - | 13 | 1634 | |||
| AK296205.1 | - | 14 | 1891 | |||
| BC033862.1 | - | 13 | 1623 | 5188554 | ||
| BX537676.1 | - | 13 | 3239 | |||
| J04813.1 | - | 13 | 1621 | |||

CYP3A5
Genomic Map