siRNA Details
| Gene Symbol: ITGA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144996.1 | NP_001138468.1 | 12 | 1938 | ||
| NM_001144997.1 | NP_001138469.1 | 12 | 1463 | |||
| NM_002206.2 | NP_002197.2 | 12 | 1926 | |||
| XM_005268839.1 | XP_005268896.1 | 13 | 2191 | |||
| XM_005268840.1 | XP_005268897.1 | 13 | 2173 | |||
| XM_005268841.1 | XP_005268898.1 | 13 | 2191 | |||
| XM_005268842.1 | XP_005268899.1 | 12 | 2041 | |||
| XM_005268844.1 | XP_005268901.1 | 14 | 1816 | |||
| XM_005268846.1 | XP_005268903.1 | 13 | 1597 | |||
| XM_005268848.1 | XP_005268905.1 | 12 | 1615 | |||
| XM_005268849.1 | XP_005268906.1 | 12 | 1545 | |||
| XM_005268850.1 | XP_005268907.1 | 10 | 1434 | |||
| XM_017019265.1 | XP_016874754.1 | 11 | 1519 | |||
| GenBank mRNA | AB210036.1 | - | 12 | 1896 | ||
| AF032108.1 | - | 12 | 1867 | |||
| AF052050.1 | - | 12 | 1706 | |||
| AF072132.1 | - | 12 | 1706 | |||
| AK022548.1 | - | 10 | 1310 | |||
| AK304864.1 | - | 12 | 1439 | |||
| AY358882.1 | - | 12 | 1737 | |||
| BC050280.1 | - | 12 | 1922 | 5284443 | ||

ITGA7
Genomic Map