siRNA Details
Gene Symbol: ITGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002206.2 | NP_002197.2 | 5 | 937 | ||
XM_005268839.1 | XP_005268896.1 | 6 | 1202 | |||
XM_005268840.1 | XP_005268897.1 | 6 | 1202 | |||
XM_005268841.1 | XP_005268898.1 | 6 | 1202 | |||
XM_005268842.1 | XP_005268899.1 | 5 | 1070 | |||
XM_005268844.1 | XP_005268901.1 | 7 | 827 | |||
XM_005268846.1 | XP_005268903.1 | 6 | 608 | |||
XM_005268848.1 | XP_005268905.1 | 5 | 626 | |||
XM_005268849.1 | XP_005268906.1 | 5 | 556 | |||
XM_005268850.1 | XP_005268907.1 | 3 | 445 | |||
XM_017019265.1 | XP_016874754.1 | 4 | 530 | |||
GenBank mRNA | AB210036.1 | - | 5 | 907 | ||
AF032108.1 | - | 5 | 878 | |||
AF052050.1 | - | 5 | 717 | |||
AF072132.1 | - | 5 | 717 | |||
AK022548.1 | - | 3 | 339 | |||
BC050280.1 | - | 5 | 933 | 5284443 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
