siRNA Details
| Gene Symbol: MYO18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078471.3 | NP_510880.2 | 2 | 997 | ||
| NM_203318.1 | NP_976063.1 | 2 | 997 | |||
| GenBank mRNA | AB177858.1 | - | 1 | 816 | ||
| BC039612.1 | - | 2 | 1101 | 6045482 | ||
| CR933614.1 | - | 2 | 972 | |||
| D86970.2 | - | 2 | 1020 | |||

MYO18A
Genomic Map