siRNA Details
| Gene Symbol: MYO18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078471.3 | NP_510880.2 | 10 | 2078 | ||
| NM_203318.1 | NP_976063.1 | 10 | 2078 | |||
| GenBank mRNA | AB177858.1 | - | 9 | 1897 | ||
| AB177860.1 | - | 10 | 1030 | |||
| AK302505.1 | - | 9 | 831 | |||
| AK303522.1 | - | 9 | 968 | |||
| AY703984.1 | - | 10 | 1030 | |||
| BC039612.1 | - | 10 | 2182 | 6045482 | ||
| CR933614.1 | - | 10 | 2053 | |||
| D86970.2 | - | 10 | 2101 | |||

MYO18A
Genomic Map