siRNA Details
| Gene Symbol: CCDC187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291516.1 | NP_001278445.1 | 2 | 412 | ||
| NR_111982.1 | 2 | 729 | ||||
| XM_011518679.2 | XP_011516981.1 | 2 | 735 | |||
| XM_011518681.2 | XP_011516983.1 | 2 | 734 | |||
| XM_011518683.2 | XP_011516985.1 | 2 | 734 | |||
| XM_011518684.2 | XP_011516986.1 | 2 | 734 | |||
| GenBank mRNA | AK125773.1 | - | 2 | 728 | ||
| BC031926.1 | - | 2 | 412 | |||
| BC042107.1 | - | 2 | 705 | 5170879 | ||

CCDC187
Genomic Map