siRNA Details
| Gene Symbol: GREB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014668.3 | NP_055483.2 | 30, 31 | 5651 | ||
| XM_005246192.4 | XP_005246249.1 | 30, 31 | 5673 | |||
| XM_005246196.3 | XP_005246253.1 | 14, 15 | 2726 | |||
| XM_011510418.2 | XP_011508720.1 | 30, 31 | 5509 | |||
| XM_011510419.2 | XP_011508721.1 | 29, 30 | 5509 | |||
| XM_011510422.2 | XP_011508724.1 | 15, 16 | 2821 | |||
| GenBank mRNA | AB011147.1 | - | 13, 14 | 2454 | ||
| BC054502.1 | - | 13, 14 | 2463 | |||

GREB1
Genomic Map