siRNA Details
| Gene Symbol: LILRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130917.2 | NP_001124389.2 | 8 | 1618 | ||
| NM_001290270.1 | NP_001277199.1 | 6 | 1531 | |||
| NM_001290271.1 | NP_001277200.1 | 9 | 2265 | |||
| NM_006866.3 | NP_006857.2 | 8 | 2139 | |||
| XM_006722986.1 | XP_006723049.1 | 9 | 2398 | |||
| XM_011526385.1 | XP_011524687.1 | 9 | 2190 | |||
| XM_011526386.1 | XP_011524688.1 | 8 | 2362 | |||
| XM_011526387.1 | XP_011524689.1 | 7 | 2103 | |||
| XM_011526389.1 | XP_011524691.1 | 7 | 2080 | |||
| XM_017026224.1 | XP_016881713.1 | 8 | 2101 | |||
| GenBank mRNA | AF025531.1 | - | 8 | 1609 | ||
| AK130170.1 | - | 7 | 1595 | |||
| AK222651.1 | - | 7 | 1563 | |||
| BC017412.1 | - | 7 | 1569 | 4618777 | ||
| BC027916.1 | - | 7 | 1552 | 5223154 | ||
| CB046242.1 | - | 3 | 525 | |||
| U82275.1 | - | 7 | 1606 | |||

LILRA2
Genomic Map