siRNA Details
Gene Symbol: LINC00200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015376.2 | 2 | 284 | |||
GenBank mRNA | AK097673.1 | - | 2 | 234 | ||
BC133038.1 | - | 2 | 284 | |||
BC133040.1 | - | 2 | 236 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015376.2 | 2 | 284 | |||
GenBank mRNA | AK097673.1 | - | 2 | 234 | ||
BC133038.1 | - | 2 | 284 | |||
BC133040.1 | - | 2 | 236 |