siRNA Details
| Gene Symbol: PNPLA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256007.2 | NP_001242936.1 | 3 | 986 | ||
| NM_001256008.2 | NP_001242937.1 | 2 | 940 | |||
| NM_001256009.2 | NP_001242938.1 | 2 | 940 | |||
| NM_001256010.2 | NP_001242939.1 | 2 | 813 | |||
| NM_001256011.2 | NP_001242940.1 | 3 | 674 | |||
| NM_015723.4 | NP_056538.1 | 4 | 1098 | |||
| XM_005250396.4 | XP_005250453.1 | 3 | 1942 | |||
| XM_011516274.2 | XP_011514576.1 | 3 | 1815 | |||
| XM_011516275.2 | XP_011514577.1 | 3 | 1815 | |||
| XM_017012261.1 | XP_016867750.1 | 2 | 1769 | |||
| GenBank mRNA | AB041261.1 | - | 2 | 766 | ||
| AF263613.1 | - | 3 | 792 | |||
| AK024335.1 | - | 3 | 674 | |||
| AK291755.1 | - | 3 | 803 | |||
| AL834147.1 | - | 2 | 693 | |||
| BC032999.2 | - | 4 | 974 | 4829433 | ||
| BX647865.1 | - | 2 | 659 | |||

PNPLA8
Genomic Map