siRNA Details
| Gene Symbol: KCTD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319294.1 | NP_001306223.1 | 13 | 1575 | ||
| NM_001319295.1 | NP_001306224.1 | 13 | 1647 | |||
| NM_016121.4 | NP_057205.2 | 13 | 1575 | |||
| XM_005273158.1 | XP_005273215.1 | 13 | 1841 | |||
| GenBank mRNA | AK055925.1 | - | 7 | 852 | ||
| AK296275.1 | - | 11 | 1108 | |||
| AK316272.1 | - | 9 | 976 | |||
| AL834389.1 | - | 7 | 799 | |||
| BC013868.2 | - | 13 | 1562 | |||
| BC094879.1 | - | 13 | 1524 | 3078971 | ||
| BC117188.1 | - | 13 | 1457 | 40125739 | ||
| BC126173.1 | - | 13 | 1457 | 8991889 | ||

KCTD3
Genomic Map