siRNA Details
| Gene Symbol: C19orf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031726.3 | NP_001026896.2 | 2 | 167 | ||
| NM_001256046.1 | NP_001242975.1 | 2 | 438 | |||
| NM_001256047.1 | NP_001242976.1 | 2 | 244 | |||
| NM_001282931.1 | NP_001269860.1 | 2 | 438 | |||
| NM_031448.4 | NP_113636.2 | 2 | 438 | |||
| GenBank mRNA | AK057185.1 | - | 2 | 135 | ||
| BC063518.1 | - | 2 | 117 | 4340755 | ||
| BG720088.1 | - | 2 | 167 | |||
| BG722048.1 | - | 2 | 406 | |||
| BI753584.1 | - | 2 | 167 | |||
| DA189248.1 | - | 2 | 244 | |||
| DA708831.1 | - | 2 | 160 | |||

C19orf12
Genomic Map