siRNA Details
| Gene Symbol: BRWD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153252.4 | NP_694984.4 | 13 | 1454 | ||
| XM_005262113.3 | XP_005262170.1 | 13 | 1456 | |||
| XM_017029384.1 | XP_016884873.1 | 9 | 984 | |||
| XM_017029385.1 | XP_016884874.1 | 13 | 1456 | |||
| GenBank mRNA | AY497046.1 | - | 13 | 1454 | ||
| AY497047.1 | - | 10 | 1386 | |||
| AY497048.1 | - | 13 | 1477 | |||
| AY497049.1 | - | 12 | 1447 | |||
| AY497050.1 | - | 13 | 1427 | |||
| AY497051.1 | - | 10 | 1014 | |||
| AY497052.1 | - | 15 | 1613 | |||
| AY497053.1 | - | 11 | 1202 | |||
| AY497054.1 | - | 10 | 1103 | |||
| AY497055.1 | - | 13 | 1388 | |||
| AY497056.1 | - | 10 | 1359 | |||
| AY497057.1 | - | 9 | 1258 | |||
| AY497058.1 | - | 9 | 1097 | |||
| AY497059.1 | - | 11 | 1468 | |||
| AY497060.1 | - | 7 | 1035 | |||
| BC111490.1 | - | 13 | 1404 | |||

BRWD3
Genomic Map