siRNA Details
| Gene Symbol: MICA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000247.2 | NP_000238.1 | 3 | 666 | ||
| NM_001177519.2 | NP_001170990.1 | 3 | 666 | |||
| NM_001289152.1 | NP_001276081.1 | 3 | 1530 | |||
| NM_001289153.1 | NP_001276082.1 | 3 | 1510 | |||
| NM_001289154.1 | NP_001276083.1 | 3 | 1338 | |||
| GenBank mRNA | AB506764.1 | - | 3 | 600 | ||
| AK299135.1 | - | 3 | 638 | |||
| AK311667.1 | - | 1 | 1529 | |||
| AY204547.1 | - | 3 | 600 | |||
| AY750850.1 | - | 3 | 600 | |||
| BC016929.1 | - | 3 | 654 | 4446575 | ||
| BM552351.1 | - | 3 | 594 | |||
| BX342160.2 | - | 3 | 624 | |||
| DT219772.1 | - | 3 | 594 | |||
| L14848.1 | - | 3 | 639 | |||

MICA
Genomic Map