siRNA Details
Gene Symbol: FOXF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001451.2 | NP_001442.2 | 2 | 1831 | ||
GenBank mRNA | BC089442.1 | - | 2 | 1820 | 30558276 | |
U13219.1 | - | 2 | 1801 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FOXF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001451.2 | NP_001442.2 | 2 | 1831 | ||
GenBank mRNA | BC089442.1 | - | 2 | 1820 | 30558276 | |
U13219.1 | - | 2 | 1801 |