siRNA Details
| Gene Symbol: FANCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004629.1 | NP_004620.1 | 10 | 1771 | ||
| GenBank mRNA | AJ007669.1 | - | 10 | 1771 | ||
| AK293427.1 | - | 11 | 1461 | |||
| AK311348.1 | - | 9 | 1245 | |||
| AK312987.1 | - | 10 | 1696 | |||
| BC000032.2 | - | 10 | 1417 | 3507258 | ||
| BC011623.2 | - | 10 | 1544 | 4053144 | ||
| BT009813.1 | - | 10 | 1279 | |||
| KU159422.1 | - | 10 | 1784 | |||
| KU178053.1 | - | 10 | 1279 | |||
| KU178054.1 | - | 10 | 1261 | |||
| KU178055.1 | - | 10 | 1279 | |||
| KU178056.1 | - | 7 | 853 | |||
| U70310.1 | - | 10 | 1413 | |||

FANCG
Genomic Map