siRNA Details
| Gene Symbol: FANCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004629.1 | NP_004620.1 | 1 | 540 | ||
| GenBank mRNA | AJ007669.1 | - | 1 | 540 | ||
| AK293427.1 | - | 2 | 230 | |||
| AK311348.1 | - | 1 | 217 | |||
| AK312987.1 | - | 1 | 465 | |||
| BC000032.2 | - | 1 | 186 | 3507258 | ||
| BC011623.2 | - | 1 | 313 | 4053144 | ||
| BT009813.1 | - | 1 | 48 | |||
| KU159422.1 | - | 1 | 571 | |||
| KU178053.1 | - | 1 | 48 | |||
| KU178054.1 | - | 1 | 48 | |||
| KU178055.1 | - | 1 | 48 | |||
| KU178056.1 | - | 1 | 48 | |||
| U70310.1 | - | 1 | 182 | |||

FANCG
Genomic Map