siRNA Details
| Gene Symbol: SERAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032861.3 | NP_116250.3 | 11 | 1253 | ||
| NR_073096.1 | 10 | 1139 | ||||
| XM_006715586.2 | XP_006715649.1 | 10 | 1182 | |||
| XM_011536196.2 | XP_011534498.1 | 10 | 1276 | |||
| XM_011536198.2 | XP_011534500.1 | 9 | 1178 | |||
| GenBank mRNA | AK027823.1 | - | 11 | 1243 | ||
| AK125878.1 | - | 12 | 2445 | |||
| BC001705.1 | - | 11 | 1210 | 3350731 | ||
| BX648104.1 | - | 10 | 1185 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA424236.1 | - | 4 | 493 | Chr. 6 - 158119149 - 158144403 [-] on Build GRCh38 | |

SERAC1
Genomic Map