siRNA Details
| Gene Symbol: NIPAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099287.1 | NP_001092757.1 | 6 | 1479 | ||
| NM_001172292.1 | NP_001165763.1 | 5 | 1422 | |||
| XM_011534552.1 | XP_011532854.1 | 6 | 1391 | |||
| GenBank mRNA | AK026158.1 | - | 6 | 1283 | ||
| AK296972.1 | - | 5 | 1422 | |||
| BC105708.1 | - | 5 | 1016 | |||
| BC105709.1 | - | 5 | 1011 | |||
| BC105710.1 | - | 5 | 1016 | |||

NIPAL4
Genomic Map