siRNA Details
| Gene Symbol: CR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006658.2 | NP_001006659.1 | 6 | 1004 | ||
| NM_001877.4 | NP_001868.2 | 6 | 1004 | |||
| XM_011509206.2 | XP_011507508.1 | 6 | 900 | |||
| GenBank mRNA | AK223627.1 | - | 6 | 976 | ||
| AK301496.1 | - | 6 | 972 | |||
| BC090937.1 | - | 6 | 939 | 3066233 | ||
| BC136394.1 | - | 6 | 978 | 9020381 | ||
| J03565.1 | - | 6 | 948 | |||
| M26004.1 | - | 6 | 954 | |||
| Y00649.1 | - | 6 | 940 | |||

CR2
Genomic Map