siRNA Details
| Gene Symbol: SLITRK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032229.2 | NP_115605.2 | 2 | 1101 | ||
| GenBank mRNA | AK021931.1 | - | 1 | 52 | ||
| AK026427.1 | - | 1 | 587 | |||
| AK292793.1 | - | 2 | 1170 | |||
| BC101070.1 | - | 2 | 1003 | 40008739 | ||
| BC101071.2 | - | 2 | 1003 | 40008742 | ||
| BC101072.2 | - | 2 | 1003 | 40008743 | ||
| BC101073.2 | - | 2 | 1003 | 40008745 | ||
| BX648640.1 | - | 2 | 947 | |||

SLITRK6
Genomic Map