siRNA Details
| Gene Symbol: LOC399886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122071.1 | 1 | 432 | |||
| GenBank mRNA | AK123417.1 | - | 1 | 433 | ||
| BC130636.1 | - | 1 | 416 | 40146667 | ||
| BC130638.1 | - | 1 | 416 | 40146669 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC399886| Gene Symbol: LOC399886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122071.1 | 1 | 432 | |||
| GenBank mRNA | AK123417.1 | - | 1 | 433 | ||
| BC130636.1 | - | 1 | 416 | 40146667 | ||
| BC130638.1 | - | 1 | 416 | 40146669 | ||