siRNA Details
| Gene Symbol: FAM180B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164379.1 | NP_001157851.1 | 3 | 435 | ||
| GenBank mRNA | BC065704.1 | - | 3 | 420 | ||
| CB999847.1 | - | 3 | 278 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM180B| Gene Symbol: FAM180B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164379.1 | NP_001157851.1 | 3 | 435 | ||
| GenBank mRNA | BC065704.1 | - | 3 | 420 | ||
| CB999847.1 | - | 3 | 278 | |||