siRNA Details
| Gene Symbol: C4orf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206981.1 | NP_001193910.1 | 3 | 641 | ||
| NM_001257072.1 | NP_001244001.1 | 3 | 462 | |||
| NM_178497.3 | NP_848592.2 | 2 | 597 | |||
| NR_046429.1 | 5 | 617 | ||||
| NR_046430.1 | 4 | 598 | ||||
| XM_011531668.2 | XP_011529970.1 | 4 | 1596 | |||
| GenBank mRNA | AK074237.1 | - | 2 | 597 | ||
| AK172776.1 | - | 2 | 597 | |||
| AK300227.1 | - | 3 | 624 | |||
| BC117342.1 | - | 2 | 596 | 40125893 | ||

C4orf26
Genomic Map