siRNA Details
| Gene Symbol: NDUFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004546.2 | NP_004537.1 | 3 | 385 | ||
| GenBank mRNA | AF050639.1 | - | 3 | 378 | ||
| AF067166.1 | - | 3 | 385 | |||
| BC001168.1 | - | 3 | 352 | |||
| BC063026.1 | - | 4 | 550 | 6064434 | ||

NDUFB2
Genomic Map